Thermo Scientific Chemicals

3-Methyl-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H52047.03
1 g, Each
Thermo Scientific Chemicals

3-Methyl-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H52047.03
1 g, Each
Quantity
Catalog number: H52047.03
also known as H52047-03
Price (USD)
Price: 114.00
Online price: 98.65
Your price:
Quantity
-

Chemical Identifiers

CAS
114926-37-3
IUPAC Name
(2S)-2-amino-3-(3-methylphenyl)propanoic acid
Molecular Formula
C10H13NO2
InChI Key
JZRBSTONIYRNRI-VIFPVBQESA-N
SMILES
CC1=CC=CC(C[C@H](N)C(O)=O)=C1
Appearance (Color)
White to pale brown
Assay (HPLC)
≥94.0%
Chiral LC
≥98% (ee)
Proton NMR
Conforms
Form
Powder

Description

It finds its application in the identification of N-acyl 4-(5-pyrimidine-2,4-dionyl)phenylalanine derivatives and their orally active prodrug esters as dual-acting alpha4-beta1 and alpha4-beta7 receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the identification of N-acyl 4-(5-pyrimidine-2,4-dionyl)phenylalanine derivatives and their orally active prodrug esters as dual-acting alpha4-beta1 and alpha4-beta7 receptor antagonists.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text