N-Fmoc-L-beta-homophenylalanine, 95%, Thermo Scientific Chemicals
N-Fmoc-L-beta-homophenylalanine, 95%, Thermo Scientific Chemicals
N-Fmoc-L-beta-homophenylalanine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Fmoc-L-beta-homophenylalanine, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H52052.03
also known as H52052-03
1 g
Catalog number H52052.03
also known as H52052-03
Price (EUR)
369,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
369,00
Each
Specifications
Beilstein7726557
CAS193954-28-8
Chemical Name or MaterialN-Fmoc-L-beta-homophenylalanine
Formula Weight401.46
IUPAC Name(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid
View more
N-Fmoc-L-beta-homophenylalanine is used for producing high quality peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-beta-homophenylalanine is used for producing high quality peptides.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Antonella Leggio; Angelo Liguori; Antonio Procopio; Giovanni Sindona. Convenient and stereospecific homologation ofN-fluorenylmethoxycarbonyl-α-amino acids totheir β-homologues. J. Chem. Soc., Perkin Trans. 1. 1997, (13),1969-1972
  2. V.V.Suresh Babu; H.N.Gopi; K.Anandi. Homologation of α-amino acids to β-amino acids using Fmoc-amino acid pentafluorophenyl esters. The Journal of Peptide Research. 1999, 53, (3),308-313