N-Fmoc-L-pentafluorophenylalanine, 95%
N-Fmoc-L-pentafluorophenylalanine, 95%
N-Fmoc-L-pentafluorophenylalanine, 95%
N-Fmoc-L-pentafluorophenylalanine, 95%
Thermo Scientific Chemicals

N-Fmoc-L-pentafluorophenylalanine, 95%

CAS: 205526-32-5 | C24H16F5NO4 | 477.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52059.06
also known as H52059-06
Price (TWD)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS205526-32-5
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid
Molecular FormulaC24H16F5NO4
InChI KeyDLOGILOIJKBYKA-KRWDZBQOSA-N
SMILESOC(=O)[C@H](CC1=C(F)C(F)=C(F)C(F)=C1F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Carbon-13 NMRConforms
Proton NMRConforms
Optical Rotation|0 = -35.2? ?2? (C=1 in DMF)
FormFused solid
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only