N-Fmoc-4-methyl-L-phenylalanine, 95%
N-Fmoc-4-methyl-L-phenylalanine, 95%
N-Fmoc-4-methyl-L-phenylalanine, 95%
Thermo Scientific Chemicals

N-Fmoc-4-methyl-L-phenylalanine, 95%

CAS: 199006-54-7 | C25H23NO4 | 401.462 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52073.03
also known as H52073-03
Price (EUR)
105,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
105,00
Each
Chemical Identifiers
CAS199006-54-7
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-methylphenyl)propanoic acid
Molecular FormulaC25H23NO4
InChI KeyUXLHLZHGQPDMJQ-QHCPKHFHSA-N
SMILESCC1=CC=C(C[C@H](NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (HPLC)≥94.0%
Water Content (Karl Fischer Titration)≤1.0%
Melting Point (clear melt)165.0-175.0?C
View more
N-Fmoc-4-methyl-L-phenylalanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-4-methyl-L-phenylalanine is used as pharmaceutical intermediate.

Solubility
Soluble in Dimethylformamide(DMF).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Gustav A.Stein; Hermann A.Bronner; K.Pfister3rd. α-Methyl α-Amino Acids. II.1 Derivatives of DL-Phenylalanine. J. Am. Chem. Soc. 1955, 77, (3),700-703