N-Fmoc-3-(3-thienyl)-L-alanine, 95%
N-Fmoc-3-(3-thienyl)-L-alanine, 95%
N-Fmoc-3-(3-thienyl)-L-alanine, 95%
Thermo Scientific Chemicals

N-Fmoc-3-(3-thienyl)-L-alanine, 95%

CAS: 186320-06-9 | C22H19NO4S | 393.457 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52167.03
also known as H52167-03
Price (EUR)
281,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
281,00
Each
Specifications
Beilstein9512976
CAS186320-06-9
Chemical Name or MaterialN-Fmoc-3-(3-thienyl)-L-alanine
Formula Weight393.46
IUPAC Name(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-3-ylpropanoic acid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only