N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El éster 6-terc-butílico de ácido N-Fmoc-L-beta-homoglutámico se utiliza como intermedio en síntesis química orgánica.
Solubilidad
Ligeramente soluble en agua.
Notas
Estable en las condiciones de almacenamiento recomendadas. Incompatible con los agentes oxidantes.
RUO – Research Use Only