Thermo Scientific Chemicals

N-Fmoc-L-beta-glutamic acid 5-tert-butyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H52190.MD
250 mg, Each
Thermo Scientific Chemicals

N-Fmoc-L-beta-glutamic acid 5-tert-butyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H52190.MD
250 mg, Each
Quantity
Catalog number: H52190.MD
also known as H52190-MD
Price (USD)
Price: 123.00
Online price: 118.65
Your price:
Quantity
-

Description

N-Fmoc-L-beta-glutamic acid 5-tert-butyl ester, is a protected building block used for the introduction of glutamic acid into peptides. Fmoc-beta-Glu(OtBu)-OH) was hydrolyzed with trifluoroacetic acid (TFA) and isolated with the HPLC system to give Fmoc-beta-Glu(OH)-OH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-beta-glutamic acid 5-tert-butyl ester, is a protected building block used for the introduction of glutamic acid into peptides. Fmoc-beta-Glu(OtBu)-OH) was hydrolyzed with trifluoroacetic acid (TFA) and isolated with the HPLC system to give Fmoc-beta-Glu(OH)-OH.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text