Azetidine-3-carboxylic acid, 98+%
Azetidine-3-carboxylic acid, 98+%
Azetidine-3-carboxylic acid, 98+%
Thermo Scientific Chemicals

Azetidine-3-carboxylic acid, 98+%

CAS: 36476-78-5 | C4H7NO2 | 101.11 g/mol
Have Questions?
Catalog NumberQuantity
H52294.03
also known as H52294-03
1 g
Catalog number H52294.03
also known as H52294-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS36476-78-5
Chemical Name or MaterialAzetidine-3-carboxylic acid
Formula Weight101.11
IUPAC Nameazetidine-3-carboxylic acid
InChI KeyGFZWHAAOIVMHOI-UHFFFAOYSA-N
View more
Azetidine-3-carboxylic acid derivatives has potential biological and foldameric applications. Azetidine-3-carboxylic acid, a fairly strained molecule provides oxadiazoles in good yields. Azetidine-3-carboxylic acid is used as a chemical hybridising agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azetidine-3-carboxylic acid derivatives has potential biological and foldameric applications. Azetidine-3-carboxylic acid, a fairly strained molecule provides oxadiazoles in good yields. Azetidine-3-carboxylic acid is used as a chemical hybridising agent

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Arthur G. Anderson,; Roger. Lok.Synthesis of azetidine-3-carboxylic acid. J. Org. Chem., 1972, 37 (24), 3953-3955.
  2. Asta Žukauskaitė,; Sven Mangelinckx,; Vida Buinauskaitė,; Algirdas Šačkus,; Norbert De Kimpe .Synthesis of new functionalized aziridine-2- and azetidine-3-carboxylic acid derivatives of potential interest for biological and foldameric applications. Amino Acids. 2011, 41 (3), 541-558.