1-Isoindolinone-5-boronic acid pinacol ester, 96%
1-Isoindolinone-5-boronic acid pinacol ester, 96%
1-Isoindolinone-5-boronic acid pinacol ester, 96%
Thermo Scientific Chemicals

1-Isoindolinone-5-boronic acid pinacol ester, 96%

CAS: 376584-62-2 | C14H18BNO3 | 259.112 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52317.03
also known as H52317-03
Price (EUR)
579,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
579,00
Each
Specifications
CAS376584-62-2
Chemical Name or Material1-Isoindolinone-5-boronic acid pinacol ester
Formula Weight259.11
IUPAC Name5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroisoindol-1-one
InChI KeyCLACMCLRELMFLJ-UHFFFAOYSA-N
View more
1-Isoindolinone-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Isoindolinone-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only