2-Fluoro-5-methoxybenzoic acid, 97+%
2-Fluoro-5-methoxybenzoic acid, 97+%
2-Fluoro-5-methoxybenzoic acid, 97+%
Thermo Scientific Chemicals

2-Fluoro-5-methoxybenzoic acid, 97+%

CAS: 367-83-9 | C8H7FO3 | 170.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H52365.03
also known as H52365-03
Price (EUR)
207,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
207,00
Each
Specifications
CAS367-83-9
Chemical Name or Material2-Fluoro-5-methoxybenzoic acid
Formula Weight170.14
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
2-Fluoro-5-methoxybenzoic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-methoxybenzoic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only
Georges Katsoulos; Sadahito Takagishi; Manfred Schlosser. The metalation of fluoroanisoles: optional regioselectivity due to metal mediated control. Synlett. 1991, 731-732