N-Boc-2-methyl-D-serine methyl ester, 97%, Thermo Scientific Chemicals
N-Boc-2-methyl-D-serine methyl ester, 97%, Thermo Scientific Chemicals
N-Boc-2-methyl-D-serine methyl ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-2-methyl-D-serine methyl ester, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H52421.03
also known as H52421-03
1 g
Catalog number H52421.03
also known as H52421-03
Price (EUR)
466,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
466,00
Each
Specifications
CAS188476-33-7
Chemical Name or MaterialN-Boc-2-methyl-D-serine methyl ester
Formula Weight233.26
IUPAC Namemethyl (2R)-3-hydroxy-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
InChI KeyOUUNEDPIBZNRMT-SNVBAGLBSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only