5-Chloropyridine-2-boronic acid, 95%
5-Chloropyridine-2-boronic acid, 95%
5-Chloropyridine-2-boronic acid, 95%
Thermo Scientific Chemicals

5-Chloropyridine-2-boronic acid, 95%

CAS: 652148-91-9 | C5H5BClNO2 | 157.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52515.03
also known as H52515-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS652148-91-9
IUPAC Name(5-chloropyridin-2-yl)boronic acid
Molecular FormulaC5H5BClNO2
InChI KeyJEGHCYRKSUGHJH-UHFFFAOYSA-N
SMILESOB(O)C1=NC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
5-Chloropyridine-2-boronic acid was used as reagent in organic synthesis. Also used as phamaceutical Intermediates. Used in suzuki-Miyaura coupling processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloropyridine-2-boronic acid was used as reagent in organic synthesis. Also used as phamaceutical Intermediates. Used in suzuki-Miyaura coupling processes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Shawn D. Walker Dr.,; Timothy E. Barder,; Joseph R. Martinelli,; Stephen L. Buchwald Prof. Dr. A Rationally Designed Universal Catalyst for Suzuki-Miyaura Coupling Processes. TOC. 20004, 43 (13), 1871-1876.