(R)-1,4-Diazabicyclo[4.3.0]nonane, 97%
(R)-1,4-Diazabicyclo[4.3.0]nonane, 97%
(R)-1,4-Diazabicyclo[4.3.0]nonane, 97%
Thermo Scientific Chemicals

(R)-1,4-Diazabicyclo[4.3.0]nonane, 97%

CAS: 96193-27-0 | C7H14N2 | 126.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52554.03
also known as H52554-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS96193-27-0
Chemical Name or Material(R)-1,4-Diazabicyclo[4.3.0]nonane
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
Formula Weight126.2
Health Hazard 1H314-H335
View more
(R)-1,4-Diazabicyclo[4.3.0]nonane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-1,4-Diazabicyclo[4.3.0]nonane is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. Meier E.Freed; Allan R.Day. Syntheses of 1,4-Diazabicyclo[4.4.0]decanes, 1,4-Diazabicyclo[4.3.0]nonanes and 1,8-Diazabicyclo[4.3.0]nonanes. J. Org. Chem. 1960, 25, (12),2108-2113
  2. Brian R. de Costa; Xiaoshu He; Joannes T.M.Linders; Celia Dominguez; Zi Qiang Gu; Wanda Williams; Wayne Bowen. Synthesis and evaluation of conformationally restricted N-[2-(3,4-dichlorophenyl)ethyl]-N-methyl-2-(1-pyrrolidinyl)ethylamines at .sigma. receptors. 2. Piperazines, bicyclic amines, bridged bicyclic amines, and miscellaneous compounds. J. Med. Chem. 1993, 36, (16),2311-2320