(R)-3-Aminomethyl-1-Boc-piperidine, 97%
(R)-3-Aminomethyl-1-Boc-piperidine, 97%
(R)-3-Aminomethyl-1-Boc-piperidine, 97%
(R)-3-Aminomethyl-1-Boc-piperidine, 97%
Thermo Scientific Chemicals

(R)-3-Aminomethyl-1-Boc-piperidine, 97%

CAS: 140645-23-4 | C11H22N2O2 | 214.309 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H52561.MD
also known as H52561-MD
Price (EUR)
48,70
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
48,70
Each
Add to cart
Chemical Identifiers
CAS140645-23-4
IUPAC Nametert-butyl 3-(aminomethyl)piperidine-1-carboxylate
Molecular FormulaC11H22N2O2
InChI KeyWPWXYQIMXTUMJB-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC(CN)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>96.0%
Residue on ignition<0.10%
Water Content (Karl Fischer Titration)<0.50%
Optical purity>98.0%
Appearance (Color)Colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only