trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride, 97%
trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride, 97%
trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride, 97%
Thermo Scientific Chemicals

trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride, 97%

CAS: 334999-32-5 | C11H21ClN2O4 | 280.75 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 H52566.06
亦稱為 H52566-06
價格 (TWD)
-
Quantity:
5 g
Request bulk or custom format
規格
CAS334999-32-5
Chemical Name or Materialtrans-4-Amino-N-Boc-L-proline methyl ester hydrochloride
Formula Weight280.75
IUPAC Name1-O-tert-butyl 2-O-methyl (2S,4R)-4-aminopyrrolidine-1,2-dicarboxylate;hydrochloride
InChI KeyKFYCQKLSGMAVQH-WLYNEOFISA-N
檢視更多
trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride acts as a pharmaceutical intermediate. It is used to prepare N-Boc-protected proline methyl ester of calix[4]arene, which is efficiently catalyzes the enantioselective aldol reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride acts as a pharmaceutical intermediate. It is used to prepare N-Boc-protected proline methyl ester of calix[4]arene, which is efficiently catalyzes the enantioselective aldol reaction.

Notes
Store in a cool place. Moisture sensitive. Incompatible with oxidizing agents and acids.
RUO – Research Use Only
Hahnenkamp, A.; Schäfers, M.; Bremer, C.; Höltke, C. Design and synthesis of small-molecule fluorescent photoprobes targeted to aminopeptdase N (APN/CD13) for optical imaging of angiogenesis. Bioconjugate Chem. 2013, 27 (6), 1027-1038.