(2S,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid, 97%
(2S,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid, 97%
(2S,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid, 97%
Thermo Scientific Chemicals

(2S,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid, 97%

CAS: 73397-27-0 | C12H23NO5 | 261.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52569.03
also known as H52569-03
Price (EUR)
558,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
558,00
Each
Specifications
CAS73397-27-0
Chemical Name or Material(2S,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid
Formula Weight261.32
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only