1-Boc-piperidine-3-carboxaldehyde, 97%
1-Boc-piperidine-3-carboxaldehyde, 97%
1-Boc-piperidine-3-carboxaldehyde, 97%
Thermo Scientific Chemicals

1-Boc-piperidine-3-carboxaldehyde, 97%

CAS: 118156-93-7 | C11H19NO3 | 213.277 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52571.03
also known as H52571-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS118156-93-7
IUPAC Nametert-butyl 3-formylpiperidine-1-carboxylate
Molecular FormulaC11H19NO3
InChI KeyCTVHINDANRPFIL-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC(C1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
FormLiquid
Assay (unspecified)>96.0%
Refractive Index1.4650-1.4700 @ 20?C
1-Boc-piperidine-3-carboxaldehyde is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-piperidine-3-carboxaldehyde is used as an organic chemical synthesis intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only