2-tert-Butyl-trans-vinylboronic acid, 97%
2-tert-Butyl-trans-vinylboronic acid, 97%
2-tert-Butyl-trans-vinylboronic acid, 97%
Thermo Scientific Chemicals

2-tert-Butyl-trans-vinylboronic acid, 97%

CAS: 86595-37-1 | C6H13BO2 | 127.978 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52627.03
also known as H52627-03
Price (EUR)
286,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
286,00
Each
Chemical Identifiers
CAS86595-37-1
IUPAC Name[(1E)-3,3-dimethylbut-1-en-1-yl]boronic acid
Molecular FormulaC6H13BO2
InChI KeyCTIWKIMYFQSVBZ-SNAWJCMRSA-N
SMILESCC(C)(C)C=CB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or crystalline powder or powder
Assay by NMR≥96.0% (1H NMR)
Proton NMRConforms to structure
2-tert-Butyl-trans-vinylboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-tert-Butyl-trans-vinylboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only