4-Methoxy-3-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
4-Methoxy-3-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
4-Methoxy-3-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxy-3-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H52660.03
also known as H52660-03
1 g
Catalog number H52660.03
also known as H52660-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS149507-36-8
IUPAC Name[4-methoxy-3-(trifluoromethyl)phenyl]boronic acid
Molecular FormulaC8H8BF3O3
InChI KeyBUSMBMGODABSIN-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1C(F)(F)F)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only