trans-N-Fmoc-4-hydroxy-L-proline, 97%
trans-N-Fmoc-4-hydroxy-L-proline, 97%
trans-N-Fmoc-4-hydroxy-L-proline, 97%
trans-N-Fmoc-4-hydroxy-L-proline, 97%
Thermo Scientific Chemicals

trans-N-Fmoc-4-hydroxy-L-proline, 97%

CAS: 88050-17-3 | C20H19NO5 | 353.374 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52740.03
also known as H52740-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS88050-17-3
IUPAC Name(2S,4R)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-4-hydroxypyrrolidine-2-carboxylic acid
Molecular FormulaC20H19NO5
InChI KeyGOUUPUICWUFXPM-XIKOKIGWSA-N
SMILESO[C@@H]1C[C@H](N(C1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to pale yellow
FormSolid
Assay (unspecified)96.0% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only