3-Fluoro-2-methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
3-Fluoro-2-methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
3-Fluoro-2-methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoro-2-methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
產品號碼Quantity
H52752.03
亦稱為 H52752-03
1 g
產品號碼 H52752.03
亦稱為 H52752-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS163517-61-1
IUPAC Name(3-fluoro-2-methylphenyl)boronic acid
Molecular FormulaC7H8BFO2
InChI KeyNYBIUWJUWTUGFV-UHFFFAOYSA-N
SMILESCC1=C(F)C=CC=C1B(O)O
檢視更多
規格Specification Sheet規格表
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only