Thermo Scientific Chemicals

Lovastatin, 97%, Thermo Scientific Chemicals

Catalog number: H52792.14
25 g, Each
Thermo Scientific Chemicals

Lovastatin, 97%, Thermo Scientific Chemicals

Catalog number: H52792.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H52792.14
also known as H52792-14
Price (USD)
Price: 317.00
Online price: 269.65
Your price:
Quantity
-

Description

Lovastatin is a cholesterol lowering drug and competitive inhibitor of HMG-CoA reductase, a rate limiting enzyme in cholesterol synthesis. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. It increases cellular resistance to anticancer agents such as doxorubicin and induces apoptosis in myeloma cells. It causes cell cycle arrest in G1 and G2/M phases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lovastatin is a cholesterol lowering drug and competitive inhibitor of HMG-CoA reductase, a rate limiting enzyme in cholesterol synthesis. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. It increases cellular resistance to anticancer agents such as doxorubicin and induces apoptosis in myeloma cells. It causes cell cycle arrest in G1 and G2/M phases.

Solubility
Soluble in DMSO (10 mg/ml), ethanol (10 mg/ml), methanol, chloroform, acetone, acetonitrile, DMF, and water (0.4 µg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text