Thermo Scientific Chemicals

4-Isocyanatobenzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: H53003.MD
250 mg, Each
Thermo Scientific Chemicals

4-Isocyanatobenzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: H53003.MD
250 mg, Each
Quantity
Catalog number: H53003.MD
also known as H53003-MD
Price (USD)
Price: 72.10
Online price: 61.65
Your price:
Quantity
-

Chemical Identifiers

CAS
380430-64-8
IUPAC Name
2-(4-isocyanatophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Formula
C13H16BNO3
InChI Key
RHKRIDVZGAXYQE-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N=C=O
Assay (HPLC)
>94.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text