規格
CAS913836-01-8
Chemical Name or Material5-(Methylsulfonyl)pyridine-3-boronic acid
Formula Weight201
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
檢視更多