Methyl 5-hexynoate, 97%
Methyl 5-hexynoate, 97%
Methyl 5-hexynoate, 97%
Thermo Scientific Chemicals

Methyl 5-hexynoate, 97%

CAS: 77758-51-1 | C7H10O2 | 126.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53370.06
also known as H53370-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Specifications
CAS77758-51-1
Chemical Name or MaterialMethyl 5-hexynoate
Formula Weight126.16
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Methyl 5-hexynoate is used in the synthesis of methyl (Z)-8-chlorooct-7-ene-5-ynoate with (Z)-1,2-dichloro-ethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-hexynoate is used in the synthesis of methyl (Z)-8-chlorooct-7-ene-5-ynoate with (Z)-1,2-dichloro-ethane.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Tang, X.; Huang, X.; Cao, T.; Han, Y.; Jiang, X.; Lin, W.; Tang, Y.; Zhang, J.; Yu, Q.; Fu, C. CuBr2-catalyzed enantioselective routes to highly functionalized and naturally occurring allenes. Org. Chem. Front. 2015, 2 (6), 688-691.
  2. Gong, J. X.; Wang, H. Y.; He, W. F.; Wang, Z. Z.; Xiao, W.; Guo, Y. W. A concise synthesis of xestospongic acid methyl ester with pancreatic lipase inhibitory activity. J. Asian Nat. Prod. Res. 2013, 15 (8), 916-919.