2-Pentynoic acid, 97%
2-Pentynoic acid, 97%
2-Pentynoic acid, 97%
Thermo Scientific Chemicals

2-Pentynoic acid, 97%

CAS: 5963-77-9 | C5H6O2 | 98.101 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H53411.14
also known as H53411-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Specifications
CAS5963-77-9
Chemical Name or Material2-Pentynoic acid
Flash Point96°C (205°F)
Formula Weight98.1
Health Hazard 1H302-H315-H319-H335
View more
2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Bruce Gomes; Gabriele Fendrich; Robert H.Abeles. Mechanism of action of glutaryl-CoA and butyryl-CoA dehydrogenases. Purification of glutaryl-CoA dehydrogenase. Biochemistry. 1981, 20, (6),1481-1490
  2. Kurt Freund; John Mizzer; William Dick; Colin Thorpe. Inactivation of general acyl-CoA dehydrogenase from pig kidney by 2-alkynoyl coenzyme A derivatives: initial aspects. Biochemistry. 1985, 24, (21),5996-6002