1-Phenyl-1-hexyn-3-ol, 97%
1-Phenyl-1-hexyn-3-ol, 97%
1-Phenyl-1-hexyn-3-ol, 97%
Thermo Scientific Chemicals

1-Phenyl-1-hexyn-3-ol, 97%

CAS: 1817-51-2 | C12H14O | 174.243 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53437.14
also known as H53437-14
Price (EUR)
263,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
263,00
Each
Specifications
CAS1817-51-2
Chemical Name or Material1-Phenyl-1-hexyn-3-ol
Density0.949
Flash Point110°C (230°F)
Formula Weight174.24
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only