3-Methyl-1-butyne is a terminal alkyne that under goes hydrosilylation reaction using phenylsilane. 3-Methyl-1-butyne is used as an intermediate for pharmaceuticals, in cosmetics and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
La 3-metil-1-butina es un alqueno terminal que se somete a una reacción de hidrosililación utilizando fenilsilano. La 3-metil-1-butina se utiliza como intermedio para los productos farmacéuticos, en cosméticos y en investigación química.
Solubilidad
Miscible con alcohol. No miscible con agua.
Notas
Almacenar en un lugar fresco. Sensible a la luz y al calor. Incompatible con ácidos, bases y agentes oxidantes fuertes.
RUO – Research Use Only
General References:
- Smolyakov, V. M.; Grebeshkov, V. V. Calculating the properties of C2H2-C9H16alkynes, based on the additivity of energy contributions. Russ. J. Phys. Chem. A 2015, 89 (5), 914-918.
- Bernal, M. J.; Martin, M.; Sola, E. Allenyl to Alkenylcarbyne Tautomerization at the Ru-Si Bond of Ru(κP,P,Si) Pincer Complexes. Organometallics 2015, 34 (4), 800-803.