1,1-Dimethylpropargylamine, 95%
1,1-Dimethylpropargylamine, 95%
1,1-Dimethylpropargylamine, 95%
Thermo Scientific Chemicals

1,1-Dimethylpropargylamine, 95%

CAS: 2978-58-7 | C5H9N | 83.134 g/mol
製品番号(カタログ番号) H53448.06
または、製品番号H53448-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2978-58-7
IUPAC Name2-methylbut-3-yn-2-amine
Molecular FormulaC5H9N
InChI KeyVUGCBIWQHSRQBZ-UHFFFAOYSA-N
SMILESCC(C)(N)C#C
さらに表示
Appearance (Color)Colorless to yellow
FormLiquid
Assay (GC)>94.0%
1,1-Dimethylpropargylamine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Dimethylpropargylamine is used as an organic chemical synthesis intermediate.

Solubility
Very soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. G.H.W.Milburn; A.Werninck; J.Tsibouklis; E.Bolton; G.Thomson; A.J.Shand. Synthesis and properties of some novel unsymmetrically substituted diacetylenes. Polymer. 1989, 30, (6),1004-1007
  2. Dipak D.Vachhani; Marzia Galli; Jeroen Jacobs; Luc Van Meervelt; Erik V.Van der Eycken. Synthesis of (spiro)cyclopentapyridinones via Csp3-H functionalization: a post-Ugi gold-catalyzed regioselective tandem cyclization. Chem. Commun. 2013, 49, (64),7171-7173