1,1-Dimethylpropargylamine, 95%
1,1-Dimethylpropargylamine, 95%
1,1-Dimethylpropargylamine, 95%
1,1-Dimethylpropargylamine, 95%
Thermo Scientific Chemicals

1,1-Dimethylpropargylamine, 95%

CAS: 2978-58-7 | C5H9N | 83.134 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53448.14
also known as H53448-14
Price (EUR)
279,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
279,00
Each
Add to cart
Chemical Identifiers
CAS2978-58-7
IUPAC Name2-methylbut-3-yn-2-amine
Molecular FormulaC5H9N
InChI KeyVUGCBIWQHSRQBZ-UHFFFAOYSA-N
SMILESCC(C)(N)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
FormLiquid
Assay (GC)>94.0%
1,1-Dimethylpropargylamine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Dimethylpropargylamine is used as an organic chemical synthesis intermediate.

Solubility
Very soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. G.H.W.Milburn; A.Werninck; J.Tsibouklis; E.Bolton; G.Thomson; A.J.Shand. Synthesis and properties of some novel unsymmetrically substituted diacetylenes. Polymer. 1989, 30, (6),1004-1007
  2. Dipak D.Vachhani; Marzia Galli; Jeroen Jacobs; Luc Van Meervelt; Erik V.Van der Eycken. Synthesis of (spiro)cyclopentapyridinones via Csp3-H functionalization: a post-Ugi gold-catalyzed regioselective tandem cyclization. Chem. Commun. 2013, 49, (64),7171-7173