4-Phenyl-3-butyn-2-ol, 97%
4-Phenyl-3-butyn-2-ol, 97%
4-Phenyl-3-butyn-2-ol, 97%
Thermo Scientific Chemicals

4-Phenyl-3-butyn-2-ol, 97%

CAS: 5876-76-6 | C10H10O | 146.189 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53450.06
also known as H53450-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Specifications
CAS5876-76-6
Chemical Name or Material4-Phenyl-3-butyn-2-ol
Density1.005
Flash Point>110°C (230°F)
Formula Weight146.19
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only