1,13-Tetradecadiene, 90+%
1,13-Tetradecadiene, 90+%
1,13-Tetradecadiene, 90+%
Thermo Scientific Chemicals

1,13-Tetradecadiene, 90+%

CAS: 21964-49-8 | C14H26 | 194.362 g/mol
Quantity:
1 g
5 g
25 g
Catalog number H53479.14
also known as H53479-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Beilstein1741459
CAS21964-49-8
Chemical Name or Material1,13-Tetradecadiene
Density0.849
EINECS Number244-687-7
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only