cis-3-Hexene, 97%
cis-3-Hexene, 97%
cis-3-Hexene, 97%
Thermo Scientific Chemicals

cis-3-Hexene, 97%

CAS: 7642-09-3 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53485.06
also known as H53485-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7642-09-3
IUPAC Name(3Z)-hex-3-ene
Molecular FormulaC6H12
InChI KeyZQDPJFUHLCOCRG-WAYWQWQTSA-N
SMILESCCC=C/CC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
Appearance (Color)Colorless
FormLiquid
cis-3-Hexene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-3-Hexene is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. L.Sjoqvist; M.Shiotani; A.Lunda. The structure of trans- and cis-3-hexene radical cations stabilised in halocarbon matrices at low temperature: An ESR and MNDO/AM1 study. Chemical Physics. 1990, 141, (2-3),417-430
  2. D. van Hemelrijk; L. van den Enden; H.J.Geise. The molecular structures of cis-3-hexene and trans-3-hexene in the gas phase by electron diffraction and molecular mechanical calculations. Journal of Molecular Structure. 1981, 74, (1),123-135