2-Bromo-4-(trifluoromethyl)phenylacetonitrile, 98%
2-Bromo-4-(trifluoromethyl)phenylacetonitrile, 98%
2-Bromo-4-(trifluoromethyl)phenylacetonitrile, 98%
Thermo Scientific Chemicals

2-Bromo-4-(trifluoromethyl)phenylacetonitrile, 98%

CAS: 474024-36-7 | C9H5BrF3N | 264.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H53518.03
also known as H53518-03
Price (EUR)
56,90
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
56,90
Each
Specifications
CAS474024-36-7
Chemical Name or Material2-Bromo-4-(trifluoromethyl)phenylacetonitrile
Density1.626
Formula Weight264.04
Health Hazard 1H302-H315-H319-H335
View more
2-Bromo-4-(trifluoromethyl)phenylacetonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-(trifluoromethyl)phenylacetonitrile is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only