5,7-Difluoro-1-indanone is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5,7-Difluoro-1-indanone is used as pharmaceutical intermediate.
Solubility
Sparingly soluble in water 0.081 g/L 25°C.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Mikhail A. Kabdulov; Konstantin Yu. Amsharov; Martin Jansen. A step toward direct fullerene synthesis: C60 fullerene precursors with fluorine in key positions. Tetrahedron. 2010, 66 (45), 8587-8593.