tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate, 97%
tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate, 97%
tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate, 97%
Thermo Scientific Chemicals

tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate, 97%

CAS: 114772-40-6 | C18H19BrO2 | 347.252 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H54100.06
also known as H54100-06
Price (EUR)
584,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
584,00
Each
Specifications
CAS114772-40-6
Chemical Name or Materialtert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate
EINECS Number442-850-9
Formula Weight347.24
Health Hazard 1H317
View more
1,1-Dimethylethyl Esterbiphenyl is used in the preparation of angiotensin II receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Dimethylethyl Esterbiphenyl is used in the preparation of angiotensin II receptor antagonists.

Solubility
Soluble in chloroform, ethyl acetate, and DMSO.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. David P. Marciano; Dana S. Kuruvilla; Siddaraju V. Boregowda; Alice Asteian; Travis S. Hughes; Ruben Garcia-Ordonez; Cesar A. Corzo; Tanya M. Khan; Scott J. Novick; HaJeung Park; Douglas J. Kojetin; Donald G. Phinney; John B. Bruning; Theodore M. Kamenecka & Patrick R. Griffin. Pharmacological repression of PPARγ promotes osteogenesis. Nature Communications. 2015, 6.
  2. Masood Hosseini; Longguang Jiang; Hans Peter Sørensen; Jan K. Jensen; Anni Christensen; Sarah Fogh; Cai Yuan; Lisbeth M. Andersen; Mingdong Huang; Peter A. Andreasen and Knud J. Jensen. Elucidation of the Contribution of Active Site and Exosite Interactions to Affinity and Specificity of Peptidylic Serine Protease Inhibitors Using Non-Natural Arginine Analogs. Molecular Pharmacology. 2009, 80 (4), 585-597.