3,4-Difluorophenylmagnesium bromide, 0.50 M in 2-MeTHF
3,4-Difluorophenylmagnesium bromide, 0.50 M in 2-MeTHF
3,4-Difluorophenylmagnesium bromide, 0.50 M in 2-MeTHF
Thermo Scientific Chemicals

3,4-Difluorophenylmagnesium bromide, 0.50 M in 2-MeTHF

CAS: 90897-92-0 | C6H3BrF2Mg | 217.30 g/mol
Have Questions?
Catalog NumberQuantity
H54197.AE
also known as H54197-AE
100 mL
Catalog number H54197.AE
also known as H54197-AE
Price (EUR)
252,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
252,00
Each
Specifications
CAS90897-92-0
Chemical Name or Material3,4-Difluorophenylmagnesium bromide
Concentration or Composition (by Analyte or Components)0.50M in 2-MeTHF
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Formula Weight217.29
View more
3,4-Difluorophenylmagnesium bromide, 0.50 M in 2-MeTHF is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Difluorophenylmagnesium bromide, 0.50 M in 2-MeTHF is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Reacts with water.
RUO – Research Use Only

General References:

  1. Yunxiang Chen; Yangwei Huang; Xueqing Zhao; & Zhongming Lia. Concise Synthesis of 1,3-Diacetoxy-2-[2'-(2″,4″-difluorophenyl)prop-2'-en-1'-yl]propane: An Intermediate for Posaconazole. Synthetic Communications: An International Journal for Rapid Communication of Synthetic Organic Chemistry. 2015, 45 (6), 734-740.
  2. Timothy A. Shepherd; James A. Aikins; David Bleakman; Buddy E. Cantrell; John P. Rearick; Richard L. Simon; Edward C. R. Smith; Gregory A. Stephenson; and Dennis M. Zimmerman. Design and Synthesis of a Novel Series of 1,2-Disubstituted Cyclopentanes as Small, Potent Potentiators of 2-Amino-3-(3-hydroxy-5-methyl-isoxazol-4-yl)propanoic Acid (AMPA) Receptors. J. Med. Chem. 2002, 45 (10), 2101-2111.