Benzofurazan-4-sulfonyl chloride, 97%
Benzofurazan-4-sulfonyl chloride, 97%
Benzofurazan-4-sulfonyl chloride, 97%
Thermo Scientific Chemicals

Benzofurazan-4-sulfonyl chloride, 97%

CAS: 114322-14-4 | C6H3ClN2O3S | 218.611 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H54275.03
also known as H54275-03
Price (EUR)
177,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
177,00
Each
Specifications
CAS114322-14-4
Chemical Name or MaterialBenzofurazan-4-sulfonyl chloride
DOT InformationHazard Class: 8; Packaging Group: III
Formula Weight218.61
Health Hazard 1H314-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only