Thermo Scientific Chemicals

O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 99+%, Thermo Scientific Chemicals

Catalog number: H54732.22
100 g, Each
Thermo Scientific Chemicals

O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 99+%, Thermo Scientific Chemicals

Catalog number: H54732.22
100 g, Each
Quantity
Catalog number: H54732.22
also known as H54732-22
Price (USD)
Price: 293.00
Online price: 261.65
Your price:
Quantity
-

Chemical Identifiers

CAS
330641-16-2
IUPAC Name
tetrafluoroboranuide; {[(6-chloro-1H-1,2,3-benzotriazol-1-yl)oxy](dimethylamino)methylidene}dimethylazanium
Molecular Formula
C11H15BClF4N5O
InChI Key
GBGVQFJZGHBZMC-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=C(Cl)C=C12)=[N+](C)C
Assay (HPLC)
>99.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text