Coumarin-6-sulfonyl chloride, 97%
Coumarin-6-sulfonyl chloride, 97%
Coumarin-6-sulfonyl chloride, 97%
Coumarin-6-sulfonyl chloride, 97%
Thermo Scientific Chemicals

Coumarin-6-sulfonyl chloride, 97%

CAS: 10543-42-7 | C9H5ClO4S | 244.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H54767.14
also known as H54767-14
Price (EUR)
638,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
638,00
Each
Add to cart
Specifications
Beilstein196167
CAS10543-42-7
Chemical Name or MaterialCoumarin-6-sulfonyl chloride
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Formula Weight244.65
View more
Coumarin-6-sulfonyl chloride is used as a derivatizing agent for amines and phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coumarin-6-sulfonyl chloride is used as a derivatizing agent for amines and phenols.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Fakhr Eldin O.Suliman; Suad S. Al-Kindi; Salma M.Z. Al-Kindy; Haider A.J. Al-Lawati. Analysis of phenols in water by high-performance liquid chromatography using coumarin-6-sulfonyl chloride as a fluorogenic precolumn label. Journal of Chromatography A. 2006, 1101,(1-2), 179-184.
  2. S.M Al-Kindy; F.E.Suliman; A.A Al-Hamadi. Fluorescence enhancement of coumarin-6-sulfonyl chloride amino acid derivatives in cyclodextrin media. Analytical Sciences. 2001, 17,(4), 539-543.