Isopropylmagnesium bromide, 3M in 2-MeTHF
Isopropylmagnesium bromide, 3M in 2-MeTHF
Isopropylmagnesium bromide, 3M in 2-MeTHF
Thermo Scientific Chemicals

Isopropylmagnesium bromide, 3M in 2-MeTHF

CAS: 920-39-8 | C3H7BrMg | 147.30 g/mol
Have Questions?
Catalog NumberQuantity
H54840.AE
also known as H54840-AE
100 mL
Catalog number H54840.AE
also known as H54840-AE
Price (EUR)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS920-39-8
IUPAC Namebromo(propan-2-yl)magnesium
Molecular FormulaC3H7BrMg
InChI KeyUCJIVFREPMUZDC-UHFFFAOYSA-M
SMILESCC(C)[Mg]Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)1.2-3.2M
CommentAqueous quench: passes test
Isopropylmagnesium bromide is a Grignard reagent used in organic synthesis, especially for the introduction of iso-propyl group to carbonyl compounds. It is also used as a strong base in aprotic solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylmagnesium bromide is a Grignard reagent used in organic synthesis, especially for the introduction of iso-propyl group to carbonyl compounds. It is also used as a strong base in aprotic solvent.

Solubility
RUO – Research Use Only

General References:

  1. Du, B. X.; Quan, Z. J.; Da, Y. X.; Zhang, Z.; Wang, X. C. Chemo-Controlled Cross-Coupling of Di(hetero)aryl Disulfides with Grignard Reagents: C-C vs. C-S Bond Formation. Adv. Synth. Catal. 2015, 357 (6), 1270-1276.
  2. Roy, A.; Bhat, B. A.; Lepore, S. D. Organo-Manganese η2-Auxiliary Directed Reactions: A Diastereoselective Approach to 2, 3-Allenols. Org. Lett. 2015, 17 (4), 900-903.