Specifications
CAS845305-87-5
Chemical Name or Material5-Bromo-N-methylpyridine-2-carboxamide
Formula Weight215.05
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more