S-Phenyl-L-cysteine, 97%
S-Phenyl-L-cysteine, 97%
S-Phenyl-L-cysteine, 97%
S-Phenyl-L-cysteine, 97%
Thermo Scientific Chemicals

S-Phenyl-L-cysteine, 97%

CAS: 34317-61-8 | C9H11NO2S | 197.252 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H55031.14
also known as H55031-14
Price (EUR)
339,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
339,00
Each
Add to cart
Specifications
Beilstein2268203
CAS34317-61-8
Chemical Name or MaterialS-Phenyl-L-cysteine
Formula Weight197.25
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only