Thermo Scientific Chemicals

Silver 2,4-pentanedionate, 98%, Thermo Scientific Chemicals

Catalog number: H55131.14
25 g, Each
Thermo Scientific Chemicals

Silver 2,4-pentanedionate, 98%, Thermo Scientific Chemicals

Catalog number: H55131.14
25 g, Each
Quantity
Catalog number: H55131.14
also known as H55131-14
Price (USD)
Price: 1,167.00
Online price: 990.65
Your price:
Quantity
-

Chemical Identifiers

CAS
15525-64-1
IUPAC Name
silver(1+) 2,4-dioxopentan-3-ide
Molecular Formula
C5H7AgO2
InChI Key
UEGHNEVWUGTMES-UHFFFAOYSA-N
SMILES
[Ag+].CC(=O)[CH-]C(C)=O
Assay (unspecified)
>97.5%

Description

Silver 2,4-pentanedionate is used to make silver impregnated nanocrystalline metal oxides, with special emphasis on aluminum oxide. It is used as a cationization reagent in combination with polar matrices such as 2,5-dihydroxybenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silver 2,4-pentanedionate is used to make silver impregnated nanocrystalline metal oxides, with special emphasis on aluminum oxide. It is used as a cationization reagent in combination with polar matrices such as 2,5-dihydroxybenzoic acid.

Solubility
Soluble in water.

Notes
Air, light and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text