Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate, Electronic grade, 99+% (metals basis)
Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate, Electronic grade, 99+% (metals basis)
Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate, Electronic grade, 99+% (metals basis)
Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate, Electronic grade, 99+% (metals basis)
Thermo Scientific Chemicals

Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate, Electronic grade, 99+% (metals basis)

CAS: 131717-99-2 | C27H33IO3S | 564.52 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 H55188.06
亦稱為 H55188-06
價格 (TWD)
-
申請報價
Quantity:
5 g
Request bulk or custom format
化學識別
CAS131717-99-2
規格Specification Sheet規格表
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Trace Metal≤10000.0ppm (≤1.0%)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only