1-Methylimidazole-2-carboxaldehyde, 98%
1-Methylimidazole-2-carboxaldehyde, 98%
1-Methylimidazole-2-carboxaldehyde, 98%
1-Methylimidazole-2-carboxaldehyde, 98%
Thermo Scientific Chemicals

1-Methylimidazole-2-carboxaldehyde, 98%

CAS: 13750-81-7 | C5H6N2O | 110.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55201.03
also known as H55201-03
Price (EUR)
46,00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
46,00
Each
Add to cart
Chemical Identifiers
CAS13750-81-7
IUPAC Name1-methyl-1H-imidazole-2-carbaldehyde
Molecular FormulaC5H6N2O
InChI KeyUEBFLTZXUXZPJO-UHFFFAOYSA-N
SMILESCN1C=CN=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water.

Notes
Air sensitive. Store in a cool, dry place. Store in a tightly closed container. Store under nitrogen.
RUO – Research Use Only

General References:

  1. Daryono H. Tjahjono.; Takehiro Akutsu.; Naoki Yoshioka.; Hidenari Inoue. Cationic porphyrins bearing diazolium rings: synthesis and their interaction with calf thymus DNA. Biochim. Biophys. Acta. 1999, 1472 (1-2),333-343.
  2. Maximilian A. Grassberger.; Friederike Turnowsky.; Johannes Hildebrandt. Preparation and antibacterial activities of new 1,2,3-diazaborine derivatives and analogs.J. Med. Chem. 1984, 27 (8), 947-953.