4-Boc-1-(5-bromo-2-pyridyl)piperazine, 97%
4-Boc-1-(5-bromo-2-pyridyl)piperazine, 97%
4-Boc-1-(5-bromo-2-pyridyl)piperazine, 97%
4-Boc-1-(5-bromo-2-pyridyl)piperazine, 97%
Thermo Scientific Chemicals

4-Boc-1-(5-bromo-2-pyridyl)piperazine, 97%

CAS: 153747-97-8 | C14H20BrN3O2 | 342.237 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
25 g
產品號碼 H55311.03
亦稱為 H55311-03
價格 (TWD)
-
申請報價
Quantity:
1 g
Request bulk or custom format
化學識別
CAS153747-97-8
IUPAC Nametert-butyl 4-(5-bromopyridin-2-yl)piperazine-1-carboxylate
Molecular FormulaC14H20BrN3O2
InChI KeyDSLVSFMWCDGZIL-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCN(CC1)C1=NC=C(Br)C=C1
檢視更多
規格Specification Sheet規格表
Assay≥96.0%
Appearance (Color)White to yellow
FormPowder or crystalline powder and/or chunks or solid
Assay (unspecified)≥96.0%
4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only