1-(1-Naphthylmethyl)piperazine, 97%
1-(1-Naphthylmethyl)piperazine, 97%
1-(1-Naphthylmethyl)piperazine, 97%
Thermo Scientific Chemicals

1-(1-Naphthylmethyl)piperazine, 97%

CAS: 40675-81-8 | C15H18N2 | 226.323 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H55398.03
also known as H55398-03
Price (EUR)
58,40
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
58,40
Each
Specifications
CAS40675-81-8
Chemical Name or Material1-(1-Naphthylmethyl)piperazine
ColorWhite to Brown
DOT InformationTransport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Formula Weight226.32
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only