Thermo Scientific Chemicals

4-Aminobiphenyl, 98%, Thermo Scientific Chemicals

Catalog number: H55424.06
5 g, Each
Thermo Scientific Chemicals

4-Aminobiphenyl, 98%, Thermo Scientific Chemicals

Catalog number: H55424.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H55424.06
also known as H55424-06
Price (USD)
Price: 65.30
Online price: 55.65
Your price:
Quantity
-

Chemical Identifiers

CAS
92-67-1
IUPAC Name
[1,1'-biphenyl]-4-amine
Molecular Formula
C12H11N
InChI Key
DMVOXQPQNTYEKQ-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=C1)C1=CC=CC=C1
Assay (HPLC)
97.5% min

Description

It is used to manufacture azo dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to manufacture azo dyes.

Solubility
Soluble in Dichloromethane, DMSO and Methanol

Notes
Store at -20°C. Avoid heat and oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text