4-(2-Pyridyl)benzaldehyde, 98%
4-(2-Pyridyl)benzaldehyde, 98%
4-(2-Pyridyl)benzaldehyde, 98%
Thermo Scientific Chemicals

4-(2-Pyridyl)benzaldehyde, 98%

CAS: 127406-56-8 | C12H9NO | 183.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55498.03
also known as H55498-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS127406-56-8
IUPAC Name4-(pyridin-2-yl)benzaldehyde
Molecular FormulaC12H9NO
InChI KeyNMLYGLCBSFKJFI-UHFFFAOYSA-N
SMILESO=CC1=CC=C(C=C1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
4-(2-Pyridyl)benzaldehyde is used as a reactive metabolite of Atazanavir

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Pyridyl)benzaldehyde is used as a reactive metabolite of Atazanavir

Solubility
Soluble in Chloroform

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Allison R. Dick.; Kami L. Hull.; Melanie S. Sanford. A Highly Selective Catalytic Method for the Oxidative Functionalization of C-H Bonds. J. Am. Chem. Soc. 2004, 126, (8), 2300-2301.
  2. Jun Han Lee.; Chang Su Lim.; Yu Shun Tian.; Ji Hee Han.; Bong Rae Cho. A Two-Photon Fluorescent Probe for Thiols in Live Cells and Tissues. J. Am. Chem. Soc. 2010, 132, (4), 1216-1217.